siRNA Details
| Gene Symbol: NKD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271082.1 | NP_001258011.1 | 7 | 605 | ||
| NM_033120.3 | NP_149111.1 | 7 | 605 | |||
| XM_005248382.3 | XP_005248439.1 | 7 | 722 | |||
| XM_005248383.3 | XP_005248440.1 | 7 | 722 | |||
| XM_011514150.2 | XP_011512452.1 | 7 | 722 | |||
| XM_017010000.1 | XP_016865489.1 | 2 | 908 | |||
| GenBank mRNA | AB062887.1 | - | 7 | 514 | ||
| AF358137.1 | - | 7 | 526 | |||
| BC004940.1 | - | 2 | 133 | |||
| BC012176.1 | - | 7 | 605 | 4649935 | ||

NKD2
Genomic Map