siRNA Details
| Gene Symbol: SAMD10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080621.4 | NP_542188.1 | 4, 5 | 769 | ||
| XM_005260199.4 | XP_005260256.1 | 6, 7 | 1906 | |||
| XM_006723705.3 | XP_006723768.1 | 5, 6 | 668 | |||
| XM_011528565.2 | XP_011526867.1 | 5, 6 | 1538 | |||
| XM_017027671.1 | XP_016883160.1 | 5, 6 | 1786 | |||
| GenBank mRNA | BC067362.1 | - | 4, 5 | 766 | 6672007 | |

SAMD10
Genomic Map