siRNA Details
| Gene Symbol: C20orf141 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256538.1 | NP_001243467.1 | 3 | 442 | ||
| NM_080739.2 | NP_542777.1 | 2 | 552 | |||
| GenBank mRNA | AK312218.1 | - | 3 | 442 | ||
| AL713708.1 | - | 2 | 530 | |||
| BC014591.1 | - | 2 | 555 | 4821474 | ||
| BX094374.1 | - | 2 | 411 | |||

C20orf141
Genomic Map