siRNA Details
| Gene Symbol: MFSD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138431.2 | NP_612440.1 | 2 | 1004 | ||
| XM_011516806.2 | XP_011515108.1 | 2 | 1951 | |||
| XM_017013005.1 | XP_016868494.1 | 2 | 1959 | |||
| GenBank mRNA | BC011982.1 | - | 2 | 871 | 3860434 | |

MFSD3
Genomic Map