siRNA Details
| Gene Symbol: KLHDC7B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138433.3 | NP_612442.2 | 1 | 1875 | ||
| GenBank mRNA | BC009980.2 | - | 1 | 1875 | 4121528 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

KLHDC7B| Gene Symbol: KLHDC7B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138433.3 | NP_612442.2 | 1 | 1875 | ||
| GenBank mRNA | BC009980.2 | - | 1 | 1875 | 4121528 | |