siRNA Details
| Gene Symbol: PRR35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145270.2 | NP_660313.1 | 2 | 353 | ||
| XM_017022959.1 | XP_016878448.1 | 2 | 563 | |||
| XM_017022960.1 | XP_016878449.1 | 3 | 1086 | |||
| XM_017022961.1 | XP_016878450.1 | 2 | 298 | |||
| GenBank mRNA | BC016613.1 | - | 2 | 353 | 3874510 | |

PRR35
Genomic Map