siRNA Details
| Gene Symbol: TRMT61A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152307.2 | NP_689520.2 | 2 | 382 | ||
| GenBank mRNA | AK097771.1 | - | 2 | 382 | ||
| BC010167.2 | - | 2 | 365 | 3960186 | ||
| DB496873.1 | - | 2 | 382 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TRMT61A| Gene Symbol: TRMT61A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152307.2 | NP_689520.2 | 2 | 382 | ||
| GenBank mRNA | AK097771.1 | - | 2 | 382 | ||
| BC010167.2 | - | 2 | 365 | 3960186 | ||
| DB496873.1 | - | 2 | 382 | |||