siRNA Details
| Gene Symbol: WBSCR27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152559.2 | NP_689772.2 | 2 | 132 | ||
| XM_017011777.1 | XP_016867266.1 | 2 | 134 | |||
| XM_017011778.1 | XP_016867267.1 | 2 | 256 | |||
| GenBank mRNA | AF534110.1 | - | 2 | 143 | ||
| AK301597.1 | - | 2 | 123 | |||
| AY354928.1 | - | 2 | 142 | |||
| BC030295.2 | - | 1 | 96 | 5396423 | ||

WBSCR27
Genomic Map