siRNA Details
Gene Symbol: CSNK1G2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033400.1 | 1 | 698 | |||
GenBank mRNA | BC029585.1 | - | 2 | 642 | 5271543 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CSNK1G2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033400.1 | 1 | 698 | |||
GenBank mRNA | BC029585.1 | - | 2 | 642 | 5271543 |