siRNA Details
Gene Symbol: NXNL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138454.1 | NP_612463.1 | 1 | 343 | ||
GenBank mRNA | BC014127.1 | - | 1 | 343 | 4579939 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: NXNL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138454.1 | NP_612463.1 | 1 | 343 | ||
GenBank mRNA | BC014127.1 | - | 1 | 343 | 4579939 |