siRNA Details
| Gene Symbol: GPHB5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF403430.1 | - | 2, 3 | 212 | ||
| BC069113.1 | - | 2, 3 | 212 | 7216930 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GPHB5| Gene Symbol: GPHB5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF403430.1 | - | 2, 3 | 212 | ||
| BC069113.1 | - | 2, 3 | 212 | 7216930 | ||