siRNA Details
| Gene Symbol: C3orf22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152533.2 | NP_689746.1 | 2, 3 | 465 | ||
| NR_130715.1 | 2, 3 | 465 | ||||
| XM_011512455.2 | XP_011510757.1 | 2, 3 | 465 | |||
| XM_011512457.2 | XP_011510759.1 | 2, 3 | 465 | |||
| XM_017005751.1 | XP_016861240.1 | 2, 3 | 435 | |||
| XM_017005752.1 | XP_016861241.1 | 2, 3 | 297 | |||
| XM_017005753.1 | XP_016861242.1 | 2, 3 | 465 | |||
| GenBank mRNA | AK097841.1 | - | 2, 3 | 465 | ||
| BC032025.1 | - | 2, 3 | 415 | 5165128 | ||
| DB055549.1 | - | 2, 3 | 465 | |||

C3orf22
Genomic Map