siRNA Details
| Gene Symbol: ZNF585A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001288800.1 | NP_001275729.1 | 5 | 1866 | ||
| NM_152655.3 | NP_689868.1 | 6 | 1968 | |||
| NM_199126.2 | NP_954577.1 | 6 | 1908 | |||
| GenBank mRNA | AK074345.1 | - | 3 | 1500 | ||
| AK290660.1 | - | 5 | 1845 | |||
| BC052956.1 | - | 6 | 1908 | |||
| BC063820.1 | - | 5 | 1800 | 6500797 | ||

ZNF585A
Genomic Map