siRNA Details
| Gene Symbol: C1orf162 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174896.3 | NP_777556.1 | 4, 5 | 378 | ||
| XM_011540712.2 | XP_011539014.1 | 5, 6 | 5858 | |||
| GenBank mRNA | BC017973.1 | - | 4, 5 | 265 | 4693393 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

C1orf162| Gene Symbol: C1orf162 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174896.3 | NP_777556.1 | 4, 5 | 378 | ||
| XM_011540712.2 | XP_011539014.1 | 5, 6 | 5858 | |||
| GenBank mRNA | BC017973.1 | - | 4, 5 | 265 | 4693393 | |