siRNA Details
| Gene Symbol: RNF151 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174903.4 | NP_777563.2 | 3 | 590 | ||
| XM_005255129.4 | XP_005255186.1 | 4 | 852 | |||
| GenBank mRNA | BC029501.2 | - | 3 | 579 | ||
| BC113014.2 | - | 3 | 590 | 40029536 | ||

RNF151
Genomic Map