siRNA Details
Gene Symbol: FAM98C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174905.3 | NP_777565.3 | 4, 5 | 570 | ||
GenBank mRNA | BC034383.1 | - | 1, 2 | 139 | ||
BC117258.1 | - | 4, 5 | 570 | 40125809 | ||
BC143830.1 | - | 4, 5 | 570 | 9052345 | ||
BI871449.1 | - | 1, 2 | 102 | |||
BM561971.1 | - | 4, 5 | 587 |

More Information
Panther Classification:
Molecular Function -




