siRNA Details
| Gene Symbol: UBAC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177967.3 | NP_808882.1 | 1 | 579 | ||
| NR_026644.1 | 1 | 579 | ||||
| XM_006719948.3 | XP_006720011.1 | 2 | 145 | |||
| XM_011521083.2 | XP_011519385.1 | 2 | 142 | |||
| GenBank mRNA | AK054563.1 | - | 1 | 580 | ||
| BC121138.1 | - | 1 | 178 | |||
| BC121139.1 | - | 1 | 178 | 40120987 | ||

UBAC2
Genomic Map