siRNA Details
| Gene Symbol: SOWAHA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175873.5 | NP_787069.4 | 1 | 1797 | ||
| GenBank mRNA | AK090823.1 | - | 1 | 224 | ||
| AK092782.1 | - | 1 | 740 | |||
| BC104771.1 | - | 1 | 926 | 8143774 | ||
| BC104775.1 | - | 1 | 926 | 8143778 | ||

SOWAHA
Genomic Map