siRNA Details
| Gene Symbol: PDE6G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002602.3 | NP_002593.1 | 3, 4 | 339 | ||
| NR_026872.1 | 2, 3 | 134 | ||||
| XM_006721942.3 | XP_006722005.1 | 3, 4 | 1312 | |||
| XM_017024734.1 | XP_016880223.1 | 3, 4 | 614 | |||
| XM_017024735.1 | XP_016880224.1 | 3, 4 | 345 | |||
| XM_017024736.1 | XP_016880225.1 | 3, 4 | 279 | |||
| GenBank mRNA | BC106884.2 | - | 3, 4 | 296 | 40029486 | |
| BQ638010.1 | - | 3, 4 | 280 | |||
| BX537414.1 | - | 3, 4 | 346 | |||
| BX647779.1 | - | 2, 3 | 128 | |||
| M36476.1 | - | 3, 4 | 296 | |||

PDE6G
Genomic Map