siRNA Details
| Gene Symbol: HIST1H2AL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003511.2 | NP_003502.1 | 1 | 335 | ||
| GenBank mRNA | BC069306.1 | - | 1 | 335 | 7262757 | |
| BC112072.1 | - | 1 | 408 | 8327540 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HIST1H2AL| Gene Symbol: HIST1H2AL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003511.2 | NP_003502.1 | 1 | 335 | ||
| GenBank mRNA | BC069306.1 | - | 1 | 335 | 7262757 | |
| BC112072.1 | - | 1 | 408 | 8327540 | ||