siRNA Details
| Gene Symbol: DNAJC25-GNG10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004125.3 | NP_004116.2 | 1, 2 | 391 | ||
| GenBank mRNA | BC015391.1 | - | 1, 2 | 357 | 4425229 | |
| BT007068.1 | - | 1, 2 | 335 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DNAJC25-GNG10| Gene Symbol: DNAJC25-GNG10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004125.3 | NP_004116.2 | 1, 2 | 391 | ||
| GenBank mRNA | BC015391.1 | - | 1, 2 | 357 | 4425229 | |
| BT007068.1 | - | 1, 2 | 335 | |||