siRNA Details
| Gene Symbol: GNG4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098721.1 | NP_001092191.1 | 3 | 402 | ||
| NM_001098722.1 | NP_001092192.1 | 3 | 528 | |||
| NM_004485.3 | NP_004476.1 | 2 | 290 | |||
| XM_006711761.2 | XP_006711824.1 | 2 | 173 | |||
| XM_011544167.2 | XP_011542469.1 | 2 | 103 | |||
| GenBank mRNA | AF493872.1 | - | 1 | 74 | ||
| AK311781.1 | - | 3 | 317 | |||
| BC022485.1 | - | 2 | 290 | 4799603 | ||
| DA122711.1 | - | 3 | 308 | |||
| DA230013.1 | - | 3 | 261 | |||
| U31382.1 | - | 2 | 172 | |||

GNG4
Genomic Map