siRNA Details
| Gene Symbol: IGFALS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146006.1 | NP_001139478.1 | 2 | 1738 | ||
| NM_004970.2 | NP_004961.1 | 2 | 1624 | |||
| NR_027389.1 | 2 | 1597 | ||||
| GenBank mRNA | AK292182.1 | - | 2 | 1591 | ||
| AK303146.1 | - | 2 | 1737 | |||
| AK313198.1 | - | 2 | 1591 | |||
| AK315808.1 | - | 2 | 1617 | |||
| BC025681.1 | - | 2 | 1624 | 5205012 | ||
| M86826.1 | - | 2 | 1599 | |||

IGFALS
Genomic Map