siRNA Details
| Gene Symbol: GNB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005273.3 | NP_005264.2 | 5 | 515 | ||
| GenBank mRNA | AF064853.1 | - | 5 | 515 | ||
| AF501883.1 | - | 4 | 242 | |||
| AK056750.1 | - | 1 | 555 | |||
| AK291592.1 | - | 5 | 390 | |||
| AL535249.3 | - | 5 | 509 | |||
| BC010073.2 | - | 5 | 439 | 4300639 | ||
| BC012348.1 | - | 5 | 500 | 4556119 | ||
| BC068003.1 | - | 5 | 425 | 6055863 | ||
| BM763069.1 | - | 5 | 512 | |||
| CR541729.1 | - | 4 | 242 | |||
| CV569909.1 | - | 5 | 391 | |||
| M16538.1 | - | 5 | 389 | |||
| M36429.1 | - | 4 | 299 | |||

GNB2
Genomic Map