siRNA Details
| Gene Symbol: CCL27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006664.3 | NP_006655.1 | 2 | 187 | ||
| GenBank mRNA | AB010445.1 | - | 2 | 187 | ||
| AF082393.1 | - | 2 | 128 | |||
| AJ243542.1 | - | 2 | 152 | |||
| BC148263.1 | - | 2 | 150 | 8860203 | ||
| BM719357.1 | - | 1 | 60 | |||

CCL27
Genomic Map