siRNA Details
| Gene Symbol: GPR182 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007264.3 | NP_009195.1 | 2 | 195 | ||
| XM_011537824.2 | XP_011536126.1 | 2 | 871 | |||
| XM_011537825.2 | XP_011536127.1 | 2 | 871 | |||
| XM_011537827.2 | XP_011536129.1 | 2 | 871 | |||
| GenBank mRNA | BC034761.1 | - | 2 | 320 | 5185932 | |
| Y13583.1 | - | 2 | 195 | |||
More Information
Set Membership: |
|

GPR182
Genomic Map