siRNA Details
| Gene Symbol: FBXO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012168.5 | NP_036300.2 | 2 | 832 | ||
| GenBank mRNA | AF174594.1 | - | 1 | 224 | ||
| AF187318.1 | - | 2 | 680 | |||
| AK308531.1 | - | 3 | 814 | |||
| AK313019.1 | - | 2 | 432 | |||
| BC025233.1 | - | 2 | 416 | 5090925 | ||
| BC096747.1 | - | 2 | 432 | 5259298 | ||
| CR542110.1 | - | 2 | 341 | |||

FBXO2
Genomic Map