siRNA Details
| Gene Symbol: PHLDA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012396.4 | NP_036528.1 | 1 | 705 | ||
| NR_073080.1 | 1 | 705 | ||||
| GenBank mRNA | AF151100.1 | - | 1 | 639 | ||
| AK075179.1 | - | 1 | 668 | |||
| AK222644.1 | - | 1 | 668 | |||
| BC014390.1 | - | 1 | 557 | 3547186 | ||
| BC068273.1 | - | 1 | 705 | 5177273 | ||
| CN356697.1 | - | 1 | 117 | |||

PHLDA3
Genomic Map