siRNA Details
| Gene Symbol: PSORS1C2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014069.2 | NP_054788.2 | 2 | 732 | ||
| GenBank mRNA | AB031480.1 | - | 2 | 723 | ||
| AF484420.1 | - | 2 | 732 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PSORS1C2| Gene Symbol: PSORS1C2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014069.2 | NP_054788.2 | 2 | 732 | ||
| GenBank mRNA | AB031480.1 | - | 2 | 723 | ||
| AF484420.1 | - | 2 | 732 | |||