siRNA Details
| Gene Symbol: KLF15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014079.3 | NP_054798.1 | 2 | 314 | ||
| XM_005247400.3 | XP_005247457.1 | 2 | 2031 | |||
| XM_011512743.2 | XP_011511045.1 | 2 | 259 | |||
| GenBank mRNA | AB029254.1 | - | 1 | 132 | ||
| BC036733.1 | - | 2 | 326 | 5526657 | ||

KLF15
Genomic Map