siRNA Details
| Gene Symbol: SURF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278928.1 | NP_001265857.1 | 2, 3 | 284 | ||
| NM_017503.4 | NP_059973.4 | 2, 3 | 284 | |||
| GenBank mRNA | BC014411.1 | - | 2, 3 | 253 | 4548425 | |
| BI118187.1 | - | 2, 3 | 226 | |||
| BM789997.1 | - | 2, 3 | 277 | |||
| BP244646.1 | - | 2, 3 | 284 | |||
| CR456753.1 | - | 2, 3 | 236 | |||
| Z35094.1 | - | 2, 3 | 253 | |||

SURF2
Genomic Map