siRNA Details
| Gene Symbol: BTBD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017797.3 | NP_060267.2 | 2, 3 | 536 | ||
| XM_005259593.1 | XP_005259650.1 | 2, 3 | 301 | |||
| GenBank mRNA | AF355797.1 | - | 2, 3 | 536 | ||
| AK225831.1 | - | 2, 3 | 535 | |||
| AY007116.1 | - | 2, 3 | 258 | |||
| BC094820.1 | - | 2, 3 | 270 | |||

BTBD2
Genomic Map