siRNA Details
| Gene Symbol: RNF126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194460.2 | NP_919442.1 | 2 | 212 | ||
| XM_005259594.1 | XP_005259651.1 | 2 | 292 | |||
| GenBank mRNA | BC001442.2 | - | 2 | 185 | 3138961 | |
| BC025374.1 | - | 2 | 139 | 4093350 | ||
| BM558139.1 | - | 283 | ||||
| BM810439.1 | - | 282 | ||||

RNF126
Genomic Map