siRNA Details
| Gene Symbol: EPN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017957.2 | NP_060427.2 | 2 | 671 | ||
| GenBank mRNA | AF324241.1 | - | 1 | 236 | ||
| AK000785.1 | - | 2 | 620 | |||
| AK291658.1 | - | 2 | 620 | |||
| AK292977.1 | - | 2 | 620 | |||
| AK295730.1 | - | 3 | 397 | |||
| AK296346.1 | - | 3 | 538 | |||
| BC001038.2 | - | 2 | 600 | |||
| BC051365.1 | - | 2 | 600 | |||
| BC077722.1 | - | 2 | 600 | |||
| BC110602.2 | - | 2 | 284 | |||
| BC110605.2 | - | 1 | 366 | 40027860 | ||

EPN3
Genomic Map