siRNA Details
| Gene Symbol: MRPL50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019051.2 | NP_061924.1 | 1 | 126 | ||
| GenBank mRNA | AK000500.1 | - | 1 | 103 | ||
| AK295521.1 | - | 1 | 93 | |||
| BC032008.1 | - | 1 | 88 | 4749199 | ||
| CN388719.1 | - | 1 | 136 | |||

MRPL50
Genomic Map