siRNA Details
| Gene Symbol: DECR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020664.3 | NP_065715.1 | 2 | 202 | ||
| GenBank mRNA | AJ293009.1 | - | 2 | 117 | ||
| AK055839.1 | - | 2 | 202 | |||
| AK128012.1 | - | 2 | 167 | |||
| AK225961.1 | - | 1 | 121 | |||
| BC010740.1 | - | 2 | 188 | 3877040 | ||

DECR2
Genomic Map