siRNA Details
| Gene Symbol: CRYGC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020989.3 | NP_066269.1 | 3 | 335 | ||
| XM_011510662.1 | XP_011508964.1 | 4 | 409 | |||
| GenBank mRNA | BC074954.2 | - | 3 | 335 | 30915279 | |
| BC074955.2 | - | 3 | 335 | 30915501 | ||
| KT455014.1 | - | 3 | 335 | |||
| U66582.1 | - | 2 | 297 | |||

CRYGC
Genomic Map