siRNA Details
| Gene Symbol: PLEKHF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024310.4 | NP_077286.3 | 2 | 401 | ||
| XM_005259256.3 | XP_005259313.1 | 2 | 689 | |||
| XM_011527309.2 | XP_011525611.1 | 2 | 1675 | |||
| GenBank mRNA | AF434818.1 | - | 1 | 299 | ||
| AK027758.1 | - | 2 | 360 | |||
| AK225814.1 | - | 2 | 633 | |||
| AK301616.1 | - | 2 | 653 | |||
| AY037145.2 | - | 2 | 765 | |||
| BC002744.2 | - | 2 | 355 | 3628796 | ||
| BC018876.2 | - | 2 | 355 | |||

PLEKHF1
Genomic Map