siRNA Details
| Gene Symbol: NUDT22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128612.2 | NP_001122084.1 | 2 | 454 | ||
| NM_001128613.2 | NP_001122085.1 | 2 | 568 | |||
| NM_001271831.1 | NP_001258760.1 | 2 | 454 | |||
| NM_032344.3 | NP_115720.1 | 2 | 568 | |||
| GenBank mRNA | AK308555.1 | - | 2 | 542 | ||
| BC006129.1 | - | 2 | 552 | 3625545 | ||
| BG705568.1 | - | 2 | 459 | |||
| BM045237.1 | - | 2 | 554 | |||
| BM923060.1 | - | 2 | 453 | |||
| HY075773.1 | - | 2 | 438 | |||

NUDT22
Genomic Map