siRNA Details
| Gene Symbol: MON1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142501.1 | NP_001135973.1 | 2 | 497 | ||
| NM_032355.3 | NP_115731.2 | 2 | 497 | |||
| XM_006713345.3 | XP_006713408.1 | 2 | 263 | |||
| XM_011534160.1 | XP_011532462.1 | 2 | 126 | |||
| GenBank mRNA | AK074404.1 | - | 2 | 307 | ||
| AK092018.1 | - | 2 | 296 | |||
| AK315630.1 | - | 2 | 351 | |||
| BC006299.2 | - | 2 | 309 | 4079816 | ||
| BC009459.1 | - | 2 | 266 | 3536968 | ||
| BC047022.1 | - | 2 | 247 | 5248424 | ||
| BQ946394.1 | - | 2 | 629 | |||

MON1A
Genomic Map