siRNA Details
| Gene Symbol: ZSCAN10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282415.1 | NP_001269344.1 | 2 | 345 | ||
| NM_032805.2 | NP_116194.2 | 3 | 808 | |||
| XM_017023791.1 | XP_016879280.1 | 2 | 844 | |||
| XM_017023792.1 | XP_016879281.1 | 2 | 844 | |||
| GenBank mRNA | AK027455.1 | - | 2 | 536 | ||
| AK074736.1 | - | 2 | 320 | |||
| BC111367.1 | - | 1 | 1137 | |||

ZSCAN10
Genomic Map