siRNA Details
| Gene Symbol: SSC4D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080744.1 | NP_542782.1 | 2 | 459 | ||
| GenBank mRNA | AJ312755.1 | - | 2 | 459 | ||
| AK172783.1 | - | 2 | 417 | |||
| BC113840.1 | - | 2 | 342 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SSC4D| Gene Symbol: SSC4D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080744.1 | NP_542782.1 | 2 | 459 | ||
| GenBank mRNA | AJ312755.1 | - | 2 | 459 | ||
| AK172783.1 | - | 2 | 417 | |||
| BC113840.1 | - | 2 | 342 | |||