siRNA Details
| Gene Symbol: XAGE5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130775.2 | NP_570131.1 | 3 | 161 | ||
| XM_017029320.1 | XP_016884809.1 | 4 | 394 | |||
| GenBank mRNA | BC069129.1 | - | 3 | 162 | 7216985 | |
| BC103962.1 | - | 2 | 459 | 40014176 | ||

XAGE5
Genomic Map