siRNA Details
| Gene Symbol: ZNF513 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001201459.1 | NP_001188388.1 | 2 | 436 | ||
| NM_144631.5 | NP_653232.3 | 3 | 513 | |||
| XM_005264142.2 | XP_005264199.1 | 3 | 346 | |||
| GenBank mRNA | AB075875.1 | - | 2 | 436 | ||
| AK096933.1 | - | 2 | 375 | |||
| AK290690.1 | - | 3 | 487 | |||
| BC052282.1 | - | 3 | 495 | 5467936 | ||
| DB024976.1 | - | 3 | 514 | |||

ZNF513
Genomic Map