siRNA Details
| Gene Symbol: IQCF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152397.2 | NP_689610.2 | 4 | 284 | ||
| XM_011533366.2 | XP_011531668.1 | 3 | 102 | |||
| XM_017005729.1 | XP_016861218.1 | 3 | 113 | |||
| GenBank mRNA | AK131018.1 | - | 4 | 235 | ||
| BC029595.1 | - | 4 | 249 | 5273825 | ||

IQCF1
Genomic Map