siRNA Details
| Gene Symbol: PROSER2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153256.3 | NP_694988.3 | 4 | 926 | ||
| XM_011519438.2 | XP_011517740.1 | 5 | 1245 | |||
| XM_017016028.1 | XP_016871517.1 | 4 | 1912 | |||
| GenBank mRNA | BC029134.1 | - | 1 | 227 | ||
| BC048800.1 | - | 4 | 926 | 5215506 | ||

PROSER2
Genomic Map