siRNA Details
| Gene Symbol: C1QC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114101.1 | NP_001107573.1 | 3 | 472 | ||
| NM_172369.3 | NP_758957.2 | 3 | 449 | |||
| GenBank mRNA | AF087892.1 | - | 3 | 447 | ||
| AK057792.1 | - | 3 | 464 | |||
| AK130613.1 | - | 3 | 447 | |||
| AK226152.1 | - | 1 | 3744 | |||
| BC009016.1 | - | 3 | 414 | 4212772 | ||
| CB995661.1 | - | 3 | 499 | |||
| DA849505.1 | - | 3 | 441 | |||

C1QC
Genomic Map