siRNA Details
| Gene Symbol: LMNTD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173573.2 | NP_775844.2 | 3 | 277 | ||
| XM_011519965.2 | XP_011518267.2 | 3 | 437 | |||
| XM_011519967.2 | XP_011518269.2 | 3 | 437 | |||
| XM_017017476.1 | XP_016872965.1 | 3 | 437 | |||
| XM_017017477.1 | XP_016872966.1 | 3 | 437 | |||
| XM_017017478.1 | XP_016872967.1 | 3 | 278 | |||
| XM_017017479.1 | XP_016872968.1 | 3 | 437 | |||
| GenBank mRNA | BC039077.1 | - | 3 | 277 | 5168905 | |
| BX384085.2 | - | 4 | 380 | |||

LMNTD2
Genomic Map