siRNA Details
| Gene Symbol: RNF207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207396.2 | NP_997279.2 | 8, 9 | 980 | ||
| XM_011541439.2 | XP_011539741.1 | 7, 8 | 1501 | |||
| XM_017001259.1 | XP_016856748.1 | 8, 9 | 934 | |||
| GenBank mRNA | AK056658.1 | - | 3, 4 | 345 | ||
| AK294223.1 | - | 3, 4 | 295 | |||
| BC034459.1 | - | 4, 5 | 815 | |||
| BC128237.1 | - | 2, 3 | 189 | |||
| DA190055.1 | - | 2, 3 | 143 | |||

RNF207
Genomic Map