siRNA Details
| Gene Symbol: ZNF707 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK095138.1 | - | 8 | 1189 | ||
| BC014894.1 | - | 7 | 565 | 3910185 | ||
| CV023324.1 | - | 3 | 284 | |||
| DA188478.1 | - | 5 | 469 | |||
| DA555314.1 | - | 6 | 537 | |||
| DA917297.1 | - | 5 | 381 | |||
| DA957023.1 | - | 6 | 492 | |||
| DA990503.1 | - | 6 | 476 | |||
| DB099015.1 | - | 6 | 537 | |||
| DB106612.1 | - | 4 | 310 | |||

ZNF707
Genomic Map